Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006801.hg.1 |
ENST00000361175.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006801.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006801.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006801.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006801.hg.1 |
TC10000621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000465996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000465996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000465996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000482258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000482258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000482258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
ENST00000482258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011346.hg.1 |
ENST00000354527.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011346.hg.1 |
ENST00000354527.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011346.hg.1 |
ENST00000354527.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011346.hg.1 |
ENST00000354527.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006800.hg.1 |
ENST00000361175.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006800.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006800.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006800.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006800.hg.1 |
TC10000621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006810.hg.1 |
ENST00000361175.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006810.hg.1 |
ENST00000371981.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006810.hg.1 |
ENST00000427144.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006810.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006810.hg.1 |
ENST00000482258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006810.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006810.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006810.hg.1 |
TC10000621.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000156584.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156584.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156584.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156584.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156584.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156584.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156584.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156584.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156584.hg.1 |
TC1000008354.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000156585.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156585.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156585.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156585.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156585.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156585.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156585.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156585.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156585.hg.1 |
TC1000008354.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000465996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000465996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000465996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000482258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000482258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000482258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
ENST00000482258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008354.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156589.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156589.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156589.hg.1 |
ENST00000465996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156589.hg.1 |
ENST00000482258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156589.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156589.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156589.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156589.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156589.hg.1 |
ENST00000465996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156589.hg.1 |
ENST00000482258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156589.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156589.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156589.hg.1 |
TC1000008354.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000156586.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156586.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156586.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156586.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156586.hg.1 |
ENST00000361175.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156586.hg.1 |
ENST00000456849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156586.hg.1 |
NM_001015880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156586.hg.1 |
NM_004670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156586.hg.1 |
TC1000008354.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011346.hg.1 |
ENST00000354527.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011346.hg.1 |
ENST00000354527.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011346.hg.1 |
ENST00000354527.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011346.hg.1 |
ENST00000354527.2 |
Ensembl |