Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000430996.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000430996.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000430996.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000430996.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000430996.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000430996.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000464917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000464917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000464917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000464917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000464917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000464917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000482265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000482265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000482265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000482265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000616661.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000616661.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000616661.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000616661.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000616661.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000616661.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000616661.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000616661.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000616661.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000616661.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000616661.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
ENST00000622213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
NM_004753.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
NM_004753.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
NM_004753.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
NM_004753.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
NM_004753.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
NM_004753.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
NM_004753.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
NM_004753.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
NM_004753.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
NM_004753.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
NM_004753.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012921.hg.1 |
NR_106788.2 |
Refseq |