Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000336984.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000336984.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000336984.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000336984.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000336984.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000336984.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000336984.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000336984.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000336984.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375104.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375104.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375104.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375104.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375104.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375104.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375104.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375104.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000375107.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000395499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000476663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000476663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000476663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000476663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000476663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000476663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000490711.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000490711.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000490711.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000490711.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000490711.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000490711.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000490711.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000490711.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
ENST00000617181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_006411.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_006411.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_006411.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_006411.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_006411.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_006411.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_006411.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_006411.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_006411.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_006411.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_032741.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_032741.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_032741.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_032741.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_032741.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_032741.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_032741.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_032741.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011481.hg.1 |
NM_032741.4 |
Refseq |