Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007692.hg.1 |
TC0X000580.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007692.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007692.hg.1 |
ENST00000535419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007692.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007693.hg.1 |
TC0X000580.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007693.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007693.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010392.hg.1 |
ENST00000582638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007690.hg.1 |
TC0X000580.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007690.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007690.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
ENST00000535419.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
ENST00000535419.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
ENST00000535419.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
ENST00000535419.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
ENST00000535419.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
NM_001282621.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
NM_001282621.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
NM_001282621.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
NM_001282621.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
NM_001282621.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008240.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156308.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156308.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156308.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156308.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156308.hg.1 |
TC0X00008240.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
ENST00000217971.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
ENST00000535419.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
ENST00000535419.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
ENST00000535419.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
ENST00000535419.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
ENST00000535419.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
NM_001282621.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
NM_001282621.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
NM_001282621.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
NM_001282621.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
NM_001282621.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008240.hg.1 |
NM_006667.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010392.hg.1 |
ENST00000582638.1 |
Ensembl |