Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011429.hg.1 |
TC04000828.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011429.hg.1 |
ENST00000261507.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011429.hg.1 |
ENST00000393766.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011429.hg.1 |
ENST00000504317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011429.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011429.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000504317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000504317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000504317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000504317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000504317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000504317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000505270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000505270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000507013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000507013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000507013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000507013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000507013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
ENST00000507013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163366.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163366.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163366.hg.1 |
ENST00000504317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163366.hg.1 |
ENST00000507013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163366.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163366.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163366.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163366.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163366.hg.1 |
ENST00000504317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163366.hg.1 |
ENST00000507013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163366.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163366.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163366.hg.1 |
TC0400009221.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000261507.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000393766.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000504317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000504317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000504317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000504317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000504317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000504317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000505270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000505270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000507013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000507013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000507013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000507013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000507013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
ENST00000507013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_001017369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009221.hg.1 |
NM_006745.4 |
Refseq |