Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005315.hg.1 |
ENST00000394389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005315.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005315.hg.1 |
TC17000448.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005322.hg.1 |
ENST00000394386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005322.hg.1 |
ENST00000394389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005322.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005322.hg.1 |
TC17000448.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005316.hg.1 |
ENST00000394389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005316.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005316.hg.1 |
TC17000448.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007677.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005321.hg.1 |
ENST00000394386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005321.hg.1 |
ENST00000394389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005321.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005321.hg.1 |
TC17000448.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005323.hg.1 |
ENST00000394386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005323.hg.1 |
ENST00000394389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005323.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005323.hg.1 |
TC17000448.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700155362.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155362.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155362.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155362.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155362.hg.1 |
TC1700007677.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700155367.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155367.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155367.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155367.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155367.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155367.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155367.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155367.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155367.hg.1 |
TC1700007677.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700155368.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155368.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155368.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155368.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155368.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155368.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155368.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155368.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155368.hg.1 |
TC1700007677.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700155360.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155360.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155360.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155360.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155360.hg.1 |
TC1700007677.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700155366.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155366.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155366.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155366.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155366.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155366.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155366.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155366.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155366.hg.1 |
TC1700007677.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000613659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000614411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007677.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155361.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155361.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155361.hg.1 |
ENST00000617516.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155361.hg.1 |
NM_007026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155361.hg.1 |
TC1700007677.hg.1 |
Clariom D TC |