Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001316.hg.1 |
TC0X000098.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001316.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001319.hg.1 |
TC0X000098.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001319.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001319.hg.1 |
ENST00000379484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001319.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001319.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001320.hg.1 |
TC0X000098.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001320.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001320.hg.1 |
ENST00000379484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001320.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001320.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000383912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000429584.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000429584.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000429584.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000429584.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000429584.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000429584.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_206923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_206923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_206923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_206923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_206923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006764.hg.1 |
NM_206923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001318.hg.1 |
TC0X000098.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001318.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001318.hg.1 |
ENST00000379484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001318.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001318.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146306.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146306.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146306.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146306.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146306.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146306.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146306.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146306.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146306.hg.1 |
TC0X00006764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00146309.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146309.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146309.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146309.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146309.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146309.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146309.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146309.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146309.hg.1 |
TC0X00006764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00146308.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146308.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146308.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146308.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146308.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146308.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146308.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146308.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146308.hg.1 |
TC0X00006764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00146304.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146304.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146304.hg.1 |
TC0X00006764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000383912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000429584.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000429584.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000429584.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000429584.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000429584.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000429584.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_206923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_206923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_206923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_206923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_206923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006764.hg.1 |
NM_206923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146307.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146307.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146307.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146307.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146307.hg.1 |
ENST00000365779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146307.hg.1 |
ENST00000379484.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146307.hg.1 |
ENST00000465888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146307.hg.1 |
NM_015884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146307.hg.1 |
TC0X00006764.hg.1 |
Clariom D TC |