Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
ENST00000275300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
ENST00000275300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
ENST00000275300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
ENST00000275300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
ENST00000275300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
ENST00000275300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
ENST00000275300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
ENST00000275300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
ENST00000275300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
ENST00000275300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
ENST00000275300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
ENST00000275300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
ENST00000275300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
NM_021977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
NM_021977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
NM_021977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
NM_021977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
NM_021977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
NM_021977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
NM_021977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
NM_021977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
NM_021977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
NM_021977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
NM_021977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010078.hg.1 |
NM_021977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013783.hg.1 |
ENST00000612111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013783.hg.1 |
ENST00000612111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013783.hg.1 |
ENST00000612111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013783.hg.1 |
ENST00000612111.1 |
Ensembl |