Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000958.hg.1 |
ENST00000380490.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000958.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000958.hg.1 |
TC13000100.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000962.hg.1 |
ENST00000380490.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000962.hg.1 |
ENST00000479597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000962.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000962.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000962.hg.1 |
TC13000100.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000964.hg.1 |
ENST00000380490.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000964.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000964.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000964.hg.1 |
TC13000100.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000479597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000479597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000479597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000960.hg.1 |
ENST00000380490.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000960.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000960.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000960.hg.1 |
TC13000100.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000957.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000957.hg.1 |
TC13000100.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300145995.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145995.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145995.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145995.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145995.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145995.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145995.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145995.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145995.hg.1 |
TC1300006770.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000479597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000479597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000479597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006770.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145993.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145993.hg.1 |
ENST00000479597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145993.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145993.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145993.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145993.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145993.hg.1 |
ENST00000479597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145993.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145993.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145993.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145993.hg.1 |
TC1300006770.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300145986.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145986.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145986.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145986.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145986.hg.1 |
TC1300006770.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300145991.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145991.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145991.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145991.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145991.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145991.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145991.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145991.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145991.hg.1 |
TC1300006770.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300145988.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145988.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145988.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145988.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145988.hg.1 |
TC1300006770.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300145987.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145987.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145987.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145987.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145987.hg.1 |
TC1300006770.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300145990.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145990.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145990.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145990.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145990.hg.1 |
ENST00000380490.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145990.hg.1 |
ENST00000617770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145990.hg.1 |
NM_001204406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145990.hg.1 |
NM_001629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145990.hg.1 |
TC1300006770.hg.1 |
Clariom D TC |