Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005660.hg.1 |
ENST00000253193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005660.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005660.hg.1 |
TC19000435.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005661.hg.1 |
ENST00000253193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005661.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005661.hg.1 |
TC19000435.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005662.hg.1 |
ENST00000253193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005662.hg.1 |
ENST00000431491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005662.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005662.hg.1 |
TC19000435.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000592484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000592484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000592484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000592484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000592484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000609744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
ENST00000609744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005659.hg.1 |
ENST00000253193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005659.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005659.hg.1 |
TC19000435.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005658.hg.1 |
ENST00000253193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005658.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005658.hg.1 |
TC19000435.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900157078.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157078.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157078.hg.1 |
TC1900007766.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900157078.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157078.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157081.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157081.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157081.hg.1 |
ENST00000592484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157081.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157081.hg.1 |
TC1900007766.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900157081.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157081.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157081.hg.1 |
ENST00000592484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157081.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000590278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000592484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000592484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000592484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000592484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000592484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000609744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
ENST00000609744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007766.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157080.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157080.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157080.hg.1 |
ENST00000592484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157080.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157080.hg.1 |
TC1900007766.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900157080.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157080.hg.1 |
ENST00000590275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157080.hg.1 |
ENST00000592484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157080.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157076.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157076.hg.1 |
NM_002333.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157076.hg.1 |
TC1900007766.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900157076.hg.1 |
ENST00000253193.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157076.hg.1 |
NM_002333.3 |
Refseq |