Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026180.hg.1 |
ENST00000262768.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026180.hg.1 |
ENST00000483343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026180.hg.1 |
ENST00000536189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026180.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026180.hg.1 |
TC17001923.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000322630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000322630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000322630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000322630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000322630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000322630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000322630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000586713.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000586713.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000586713.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000586713.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000586713.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000586713.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000587052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000587052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000587052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000587052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000587052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000587052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000590267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000590267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000590267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000590267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000590267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000590267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000619342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000619342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000619342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
ENST00000619342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_001243540.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_001243540.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_001243540.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_001243540.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_001243540.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_001243540.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_001243540.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000322630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000322630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000322630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000322630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000322630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000322630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000322630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000586713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000586713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000586713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000586713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000586713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000586713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000587052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000587052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000587052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000587052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000587052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000587052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000590267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000590267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000590267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000590267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000590267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000590267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000619342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000619342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000619342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
ENST00000619342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_001243540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_001243540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_001243540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_001243540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_001243540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_001243540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_001243540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_001243541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011919.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197353.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197353.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197353.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197353.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197353.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197353.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197353.hg.1 |
ENST00000262768.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197353.hg.1 |
ENST00000536189.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197353.hg.1 |
ENST00000585421.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197353.hg.1 |
ENST00000586057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197353.hg.1 |
ENST00000592761.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197353.hg.1 |
NM_003255.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197353.hg.1 |
TC1700011919.hg.1 |
Clariom D TC |