Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000241393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000241393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000241393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000241393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000241393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000241393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000241393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000241393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000241393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000409817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000409817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000409817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000409817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000409817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000409817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000409817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000409817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000409817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000466288.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000466288.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000466288.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000466288.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000466288.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000466288.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
ENST00000466288.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_001008540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_001008540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_001008540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_001008540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_001008540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_001008540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_001008540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_001008540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_001008540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_003467.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_003467.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_003467.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_003467.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_003467.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_003467.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_003467.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_003467.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014414.hg.1 |
NM_003467.2 |
Refseq |