Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003481.hg.1 |
TC6_ssto_hap7000213.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003481.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003481.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018764.hg.1 |
TC06004126.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018764.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018764.hg.1 |
ENST00000374899.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018764.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018764.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001715.hg.1 |
TC6_apd_hap1000130.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001715.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001715.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003660.hg.1 |
TC6_dbb_hap3000242.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003660.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003660.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003722.hg.1 |
TC6_qbl_hap6000185.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003722.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003722.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003012.hg.1 |
TC6_mann_hap4000165.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003012.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003012.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018763.hg.1 |
TC06004126.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018763.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018763.hg.1 |
ENST00000374899.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018763.hg.1 |
ENST00000485701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018763.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018763.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003661.hg.1 |
TC6_dbb_hap3000242.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003661.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003661.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001716.hg.1 |
TC6_apd_hap1000130.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001716.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001716.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003547.hg.1 |
TC6_mcf_hap5000225.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003547.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003547.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000464100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000464100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000464100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000464100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000464100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000464100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000485701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000485701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000485701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003907.hg.1 |
TC6_cox_hap2000257.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003907.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003907.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003908.hg.1 |
TC6_cox_hap2000257.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003908.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003908.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003011.hg.1 |
TC6_mann_hap4000165.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003011.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003011.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003548.hg.1 |
TC6_mcf_hap5000225.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003548.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003548.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003723.hg.1 |
TC6_qbl_hap6000185.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003723.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003723.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003482.hg.1 |
TC6_ssto_hap7000213.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003482.hg.1 |
NM_000544.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003482.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600201516.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201516.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201516.hg.1 |
ENST00000485701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201516.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201516.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600201516.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201516.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201516.hg.1 |
ENST00000485701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201516.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201516.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600201516.hg.1 |
TC0600014275.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600201517.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201517.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201517.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201517.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600201517.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201517.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201517.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600201517.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600201517.hg.1 |
TC0600014275.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374897.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000374899.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000464100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000464100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000464100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000464100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000464100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000464100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000485701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000485701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000485701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
ENST00000620123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014275.hg.1 |
NM_018833.2 |
Refseq |