Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005157.5 | NP_005148.2 | 3 - 4 | 738 | 91 | |
NM_007313.2 | NP_009297.2 | 3 - 4 | 1042 | 91 | ||
GenBank mRNA | AB209456.1 | - | 3 - 4 | 748 | 91 | |
AB209642.1 | - | 3 - 4 | 971 | 91 | ||
BC117451.1 | - | 3 - 4 | 668 | 40126002 | 91 | |
M14752.1 | - | 3 - 4 | 910 | 91 | ||
X16416.1 | - | 3 - 4 | 693 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Endogenous Controls Homo sapiens HUMAN ENDOGENOUS CONTROL FAST PLATE Inventoried Human_Endogenous_Controls Probe spans exons |