Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000473564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000473564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000473564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000473564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000495499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000495499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000495499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000495499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000495499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000497741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000497741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000497741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000497741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000497741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000497741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
ENST00000497741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001059.hg.1 |
TC07000065.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001059.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001059.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000348035.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000473564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000473564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000473564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000473564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000488373.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000495499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000495499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000495499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000495499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000495499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000497741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000497741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000497741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000497741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000497741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000497741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
ENST00000497741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_006908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006648.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700145838.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145838.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700145838.hg.1 |
ENST00000356142.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145838.hg.1 |
NM_018890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700145838.hg.1 |
TC0700006648.hg.1 |
Clariom D TC |