Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000518991.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000518991.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000518991.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000518991.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000614426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000614426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000614426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000614426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000614426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
ENST00000614426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
NM_001277971.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
NM_001277971.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
NM_001277971.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
NM_001277971.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
NM_001277971.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
NM_001277971.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007459.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004887.hg.1 |
TC08000332.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004887.hg.1 |
ENST00000331373.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004887.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000518991.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000518991.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000518991.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000518991.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000614426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000614426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000614426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000614426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000614426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
ENST00000614426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
NM_001277971.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
NM_001277971.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
NM_001277971.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
NM_001277971.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
NM_001277971.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
NM_001277971.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007459.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151714.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151714.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151714.hg.1 |
ENST00000331373.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151714.hg.1 |
NM_032237.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151714.hg.1 |
TC0800007459.hg.1 |
Clariom D TC |