Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060800.hg.1 |
TC01004025.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060800.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060800.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000493477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000493477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000493477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000497042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000497042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
ENST00000497042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000493477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000493477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000493477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000497042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000497042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
ENST00000497042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017945.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100235149.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100235149.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100235149.hg.1 |
ENST00000366560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100235149.hg.1 |
NM_000143.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100235149.hg.1 |
TC0100017945.hg.1 |
Clariom D TC |