Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000537057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000537057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000537057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000537057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000537400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000537400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000537400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000537400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000538271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000538271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000545223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000545223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000545223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000545223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000545223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000545223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000545870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000545870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000545870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000545870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000545870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000630317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
ENST00000630317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012138.hg.1 |
ENST00000229328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012138.hg.1 |
ENST00000539596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012138.hg.1 |
ENST00000540121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012138.hg.1 |
ENST00000541640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012138.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012138.hg.1 |
TC12000936.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200167267.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167267.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167267.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167267.hg.1 |
ENST00000545223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167267.hg.1 |
ENST00000545870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167267.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167267.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167267.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167267.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167267.hg.1 |
ENST00000545223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167267.hg.1 |
ENST00000545870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167267.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167267.hg.1 |
TC1200009103.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000229328.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000537057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000537057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000537057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000537057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000537400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000537400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000537400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000537400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000538271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000538271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000540121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000541640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000542698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000545223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000545223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000545223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000545223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000545223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000545223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000545870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000545870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000545870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000545870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000545870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000630317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
ENST00000630317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009103.hg.1 |
NM_006253.4 |
Refseq |