Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000328735.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000330333.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000347667.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000463674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000463674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000463674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000463674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000463674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000463674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000463674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000463674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000463674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000463674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000465326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000465326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000465326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000465326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000465326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000465326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000465326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000465326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000466122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000466122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000466122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000466122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000466122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000466122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000466122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000466122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000466122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000466122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000466122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000466122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000466122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000470864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000470864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000470864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000470864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000475618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000475618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000487994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000487994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000487994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000487994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000487994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000487994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000487994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000487994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000487994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000491838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000491838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000491838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000491838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000491838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000491838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
ENST00000491838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_012105.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138991.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007198.hg.1 |
NM_138992.2 |
Refseq |