Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000320996.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000320996.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000320996.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000320996.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000320996.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000320996.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000320996.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000320996.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000320996.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000320996.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000320996.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000320996.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000320996.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000320996.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000320996.14 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000335272.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000415296.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000415296.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000415296.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000415296.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000415296.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000415296.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000415296.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000415296.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000415296.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000436663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000436663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000436663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000436663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000436663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000436663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000436663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000455418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000460566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000460566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000460566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000460566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000465179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000465179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000465179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000465179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000471825.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000471825.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000471825.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000477861.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000477861.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000477861.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000477861.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000477861.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000477861.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000477861.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000477861.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000477861.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000477861.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000477861.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000493007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000493007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000493007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000493007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
ENST00000634017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_001284278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008350.hg.1 |
NM_012399.4 |
Refseq |