Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022198.hg.1 |
TC01001421.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022198.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022198.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022198.hg.1 |
ENST00000538489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022198.hg.1 |
ENST00000545294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022198.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022198.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022198.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010379.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176472.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176472.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176472.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176472.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176472.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176472.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176472.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176472.hg.1 |
ENST00000282169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176472.hg.1 |
ENST00000489294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176472.hg.1 |
ENST00000538489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176472.hg.1 |
ENST00000545294.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176472.hg.1 |
NM_001184763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176472.hg.1 |
NM_144624.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176472.hg.1 |
NM_175866.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176472.hg.1 |
TC0100010379.hg.1 |
Clariom D TC |