Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008340.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008338.hg.1 |
ENST00000542477.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008338.hg.1 |
ENST00000542477.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008338.hg.1 |
ENST00000542477.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008338.hg.1 |
ENST00000542477.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008338.hg.1 |
ENST00000544535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008338.hg.1 |
ENST00000544535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008338.hg.1 |
ENST00000544535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009799.hg.1 |
ENST00000312293.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009799.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009799.hg.1 |
ENST00000393679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009799.hg.1 |
ENST00000393681.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009799.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009799.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009799.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009799.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009799.hg.1 |
TC11000754.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008340.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008338.hg.1 |
ENST00000542477.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008338.hg.1 |
ENST00000542477.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008338.hg.1 |
ENST00000542477.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008338.hg.1 |
ENST00000542477.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008338.hg.1 |
ENST00000544535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008338.hg.1 |
ENST00000544535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008338.hg.1 |
ENST00000544535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
ENST00000312293.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
ENST00000393676.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
ENST00000393679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
ENST00000393681.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
NM_000802.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
NM_016724.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
NM_016725.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
NM_016729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100161660.hg.1 |
TC1100008340.hg.1 |
Clariom D TC |