Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060750 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060749 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060732 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060769 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060734 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060757 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074870 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074887 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060719 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060712 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060739 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060731 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060707 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060730 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060770 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060744 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060705 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060725 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15029117 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060751 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074828 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060761 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060768 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074818 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060723 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074819 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060767 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060717 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074824 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060737 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074817 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074825 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074820 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074823 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074882 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060745 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060755 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074874 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060729 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060721 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060706 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074869 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060748 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060756 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060747 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060743 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060722 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060736 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074888 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060727 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074850 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074830 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060711 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00060760 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20074879 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
90014 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
34270 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
240411 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
288667 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
882758 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
504021 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
876408 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P103720 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P103720 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_24_P936171 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
32994 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
31424 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P104862 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P104862 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
25496 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
47518 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
54119 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
62588 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
57599 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
4819 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
30807 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
33247 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
52571 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
28697 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P00019551 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P30048780 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P00019556 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P30048789 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P00019560 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P00019553 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P00019547 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P103720 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_23_P103720 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074881 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060751 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074869 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060756 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060748 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060757 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074868 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060769 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15029117 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074882 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074857 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074879 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060750 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074887 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074874 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060760 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060744 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060761 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060742 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060767 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060747 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074880 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060755 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060748 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074881 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060757 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074879 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15029117 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074869 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060750 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074882 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060747 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060767 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060755 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074887 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074868 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060769 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060761 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060751 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074874 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060756 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074857 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060744 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060742 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00060760 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20074880 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_24_P936171 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P103720 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P104862 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00060752 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00060769 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00060747 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00060771 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00060763 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00060742 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00060761 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00060748 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00060767 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00060762 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00060751 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00060756 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00060758 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00060753 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00060765 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00060750 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15029121 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00060751 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15029122 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15029120 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15029124 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15029123 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00060749 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00060748 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P104862 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P00019552 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P104862 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P103720 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_24_P936171 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
154410 |
AK223482.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
154410 |
AK314791.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
154410 |
BC002518.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
154410 |
BC007585.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
154410 |
BC053555.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
154410 |
BX647151.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
154410 |
CR600287.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
154410 |
NM_002466.2 |
refseq_mrna_human_NM |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
154410 |
X13293.1 |
genbank_mrna_human |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
19039 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P10037876 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P15037843 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P30048806 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P10038149 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P104862 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00019552 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060748 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060767 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060762 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060736 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060711 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060765 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060771 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060742 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15029122 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060701 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060720 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060713 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060727 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060756 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060716 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060752 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060769 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060710 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060761 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060745 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060726 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060729 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060763 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060758 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060747 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060734 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060739 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060718 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060732 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060704 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060723 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060706 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060753 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060751 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15029117 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060708 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060740 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00060722 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P104862 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P00019552 |
- |
- |