Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014315.hg.1 |
ENST00000444815.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015781.hg.1 |
TC09001228.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015781.hg.1 |
ENST00000286548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015781.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000411677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000411677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000411677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
ENST00000411677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015778.hg.1 |
TC09001228.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015778.hg.1 |
ENST00000286548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015778.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015774.hg.1 |
TC09001228.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015774.hg.1 |
ENST00000286548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015774.hg.1 |
ENST00000397476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015774.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015776.hg.1 |
TC09001228.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015776.hg.1 |
ENST00000286548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015776.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015773.hg.1 |
TC09001228.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015773.hg.1 |
ENST00000286548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015773.hg.1 |
ENST00000397476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015773.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171746.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171746.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171746.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171746.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171746.hg.1 |
TC0900010485.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900171737.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171737.hg.1 |
ENST00000411677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171737.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171737.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171737.hg.1 |
ENST00000411677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171737.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171737.hg.1 |
TC0900010485.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900171739.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171739.hg.1 |
ENST00000411677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171739.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171739.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171739.hg.1 |
ENST00000411677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171739.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171739.hg.1 |
TC0900010485.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014315.hg.1 |
ENST00000444815.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171742.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171742.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171742.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171742.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171742.hg.1 |
TC0900010485.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900171747.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171747.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171747.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900171747.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900171747.hg.1 |
TC0900010485.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000286548.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000411677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000411677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000411677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
ENST00000411677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010485.hg.1 |
NM_002072.4 |
Refseq |