Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000354258.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000354258.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000354258.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000354258.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000354258.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000354258.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000354258.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000354258.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000354258.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000354258.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000354258.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000354258.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000354258.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000354258.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000486332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000486332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000486332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000486332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000486332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000486332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000486332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000486332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000486332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000486332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000487296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000487296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000487296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000487296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000487296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
ENST00000487296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
NM_001292022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
NM_001292022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
NM_001292022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
NM_001292022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
NM_001292022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
NM_001292022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
NM_001292022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
NM_001292022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
NM_001292022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
NM_001292022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
NM_001292022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
NM_001292022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011508.hg.1 |
NM_001292022.1 |
Refseq |