Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014504.hg.1 |
TC06001220.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014504.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014504.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014506.hg.1 |
TC06001220.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014506.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014506.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014507.hg.1 |
TC06001220.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014507.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014507.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200232.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200232.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200232.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200232.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200232.hg.1 |
TC0600014220.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014220.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200231.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200231.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200231.hg.1 |
ENST00000380698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200231.hg.1 |
NM_004155.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200231.hg.1 |
TC0600014220.hg.1 |
Clariom D TC |