Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000413826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000413826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000413826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019255.hg.1 |
TC01001262.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019255.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019255.hg.1 |
ENST00000428723.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019255.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019256.hg.1 |
TC01001262.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019256.hg.1 |
ENST00000368677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019256.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019256.hg.1 |
ENST00000428723.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019256.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000413826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000413826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000413826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172822.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172822.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172822.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172822.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172822.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172822.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172822.hg.1 |
TC0100010047.hg.1 |
Clariom D TC |