Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024320.hg.1 |
TC05003406.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024320.hg.1 |
ENST00000481195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024320.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000231504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000231504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000231504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000231504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000472253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000472253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000472253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000472253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000495833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000495833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000495833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000522385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000522385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000522385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000522385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000523082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000523082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
ENST00000523082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196633.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196633.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196633.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196633.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196633.hg.1 |
TC0500013359.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000231504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000231504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000231504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000231504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000472253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000472253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000472253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000472253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000481195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000495833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000495833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000495833.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000522385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000522385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000522385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000522385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000523082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000523082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
ENST00000523082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013359.hg.1 |
NM_002715.3 |
Refseq |