Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003672.hg.1 |
TC6_dbb_hap3000184.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003672.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003672.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003676.hg.1 |
TC6_dbb_hap3000184.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003676.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003676.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003559.hg.1 |
TC6_mcf_hap5000173.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003559.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003559.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003563.hg.1 |
TC6_mcf_hap5000173.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003563.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5003563.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003926.hg.1 |
TC6_cox_hap2000198.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003926.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003926.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003499.hg.1 |
TC6_ssto_hap7000168.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003499.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003499.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018776.hg.1 |
TC06001573.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018776.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018776.hg.1 |
ENST00000374882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018776.hg.1 |
ENST00000395339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018776.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018776.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003734.hg.1 |
TC6_qbl_hap6000186.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003734.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003734.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001731.hg.1 |
TC6_apd_hap1000104.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001731.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001731.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001733.hg.1 |
TC6_apd_hap1000104.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001733.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001733.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003738.hg.1 |
TC6_qbl_hap6000186.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003738.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003738.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003740.hg.1 |
TC6_qbl_hap6000186.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003740.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6003740.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003497.hg.1 |
TC6_ssto_hap7000168.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003497.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003497.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000490613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000490613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000490613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000490613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
ENST00000490613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018780.hg.1 |
TC06001573.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018780.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018780.hg.1 |
ENST00000374882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018780.hg.1 |
ENST00000395339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018780.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018780.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003924.hg.1 |
TC6_cox_hap2000198.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003924.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003924.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003678.hg.1 |
TC6_dbb_hap3000184.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003678.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003678.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003027.hg.1 |
TC6_mann_hap4000166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003027.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003027.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018782.hg.1 |
TC06001573.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018782.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018782.hg.1 |
ENST00000374882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018782.hg.1 |
ENST00000395339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018782.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06018782.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003920.hg.1 |
TC6_cox_hap2000198.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003920.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003920.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003029.hg.1 |
TC6_mann_hap4000166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003029.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003029.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003493.hg.1 |
TC6_ssto_hap7000168.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003493.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7003493.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001727.hg.1 |
TC6_apd_hap1000104.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001727.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001727.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003023.hg.1 |
TC6_mann_hap4000166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003023.hg.1 |
NM_004159.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4003023.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600178570.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178570.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178570.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178570.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178570.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600178570.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178570.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178570.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178570.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178570.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600178570.hg.1 |
TC0600011507.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600178571.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178571.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178571.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178571.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178571.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600178571.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178571.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178571.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178571.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178571.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600178571.hg.1 |
TC0600011507.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000490613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000490613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000490613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000490613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
ENST00000490613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011507.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600178568.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178568.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178568.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178568.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178568.hg.1 |
ENST00000490613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178568.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600178568.hg.1 |
ENST00000374881.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178568.hg.1 |
ENST00000374882.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178568.hg.1 |
ENST00000395339.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178568.hg.1 |
ENST00000484003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178568.hg.1 |
ENST00000490613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178568.hg.1 |
NM_148919.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600178568.hg.1 |
TC0600011507.hg.1 |
Clariom D TC |