Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026151.hg.1 |
TC03001445.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026151.hg.1 |
ENST00000296483.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026151.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026151.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
ENST00000469623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
ENST00000469623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
ENST00000469623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026149.hg.1 |
TC03001445.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026149.hg.1 |
ENST00000296483.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026149.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026149.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026152.hg.1 |
TC03001445.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026152.hg.1 |
ENST00000296483.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026150.hg.1 |
TC03001445.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026150.hg.1 |
ENST00000296483.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026150.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026150.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183207.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183207.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183207.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183207.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183207.hg.1 |
TC0300011152.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
ENST00000469623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
ENST00000469623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
ENST00000469623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011152.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183206.hg.1 |
ENST00000469623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183206.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183206.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183206.hg.1 |
ENST00000469623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183206.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183206.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183206.hg.1 |
TC0300011152.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300183208.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183208.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183208.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183208.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183208.hg.1 |
TC0300011152.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300183209.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183209.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183209.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183209.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183209.hg.1 |
TC0300011152.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300183205.hg.1 |
ENST00000469623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183205.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183205.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183205.hg.1 |
ENST00000469623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183205.hg.1 |
ENST00000495880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183205.hg.1 |
NM_001947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183205.hg.1 |
TC0300011152.hg.1 |
Clariom D TC |