Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000436216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000436216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000436216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000436216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000436216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020394.hg.1 |
TC08001548.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020394.hg.1 |
ENST00000378204.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020394.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000436216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000436216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000436216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000436216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000436216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011579.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178672.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178672.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178672.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178672.hg.1 |
ENST00000378204.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178672.hg.1 |
ENST00000437196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178672.hg.1 |
NM_000127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178672.hg.1 |
TC0800011579.hg.1 |
Clariom D TC |