Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000374994.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000546584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000546584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000546584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000547314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000547314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000547314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000547314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000548365.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000548365.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000548365.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000548365.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000549766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000550253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000550253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000550253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000550253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000550253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000550253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000550253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000550253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000550253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000550253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000550253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552573.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552573.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
ENST00000552573.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001130916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001130916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001130916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001130916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001130916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001130916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001130916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001130916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001130916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001130916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001130916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001130916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001130916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001130916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001130916.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_001306210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008202.hg.1 |
NM_004612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009436.hg.1 |
ENST00000594355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009436.hg.1 |
ENST00000594355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009436.hg.1 |
ENST00000594355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009436.hg.1 |
ENST00000594355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009436.hg.1 |
ENST00000594355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009436.hg.1 |
ENST00000594355.1 |
Ensembl |