Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014893.hg.1 |
TC03000835.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014893.hg.1 |
ENST00000305097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014893.hg.1 |
NM_002563.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009224.hg.1 |
ENST00000305097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009224.hg.1 |
ENST00000305097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009224.hg.1 |
ENST00000305097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009224.hg.1 |
ENST00000305097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009224.hg.1 |
ENST00000305097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009224.hg.1 |
ENST00000305097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009224.hg.1 |
ENST00000460407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009224.hg.1 |
NM_002563.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009224.hg.1 |
NM_002563.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009224.hg.1 |
NM_002563.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009224.hg.1 |
ENST00000305097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009224.hg.1 |
ENST00000305097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009224.hg.1 |
ENST00000305097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009224.hg.1 |
ENST00000305097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009224.hg.1 |
ENST00000305097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009224.hg.1 |
ENST00000305097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009224.hg.1 |
ENST00000460407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009224.hg.1 |
NM_002563.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009224.hg.1 |
NM_002563.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009224.hg.1 |
NM_002563.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166136.hg.1 |
ENST00000305097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166136.hg.1 |
NM_002563.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166136.hg.1 |
ENST00000305097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166136.hg.1 |
NM_002563.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166136.hg.1 |
TC0300009224.hg.1 |
Clariom D TC |