Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039366.1 | NP_001034455.1 | 8 | 1008 | 97 | |
NM_001039367.1 | NP_001034456.1 | 8 | 984 | 97 | ||
NM_001696.3 | NP_001687.1 | 9 | 1074 | 97 | ||
GenBank mRNA | AI565049.1 | - | 1 | 97 | ||
AK294623.1 | - | 8 | 935 | 97 | ||
AK315941.1 | - | 8 | 910 | 97 | ||
AW406069.1 | - | 2 | 301 | 97 | ||
BC004443.1 | - | 9 | 976 | 2820930 | 97 | |
X71491.1 | - | 9 | 943 | 97 | ||
X76228.1 | - | 9 | 962 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon |