Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000460015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000460015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000463191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000463191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000463191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000463191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000463191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000465318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000465318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000465318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000465318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000465318.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000467888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000467888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000467888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000467888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000467888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000468530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000468530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000468530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000468530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000468530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000468530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000475822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000475822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000475822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000475822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000483692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000483692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000483692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000483692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
ENST00000483692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004267.hg.1 |
TC02000281.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004267.hg.1 |
ENST00000263734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004267.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151626.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151626.hg.1 |
ENST00000483692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151626.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151626.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151626.hg.1 |
ENST00000483692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151626.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151626.hg.1 |
TC0200007458.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000263734.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000449347.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000460015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000460015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000463191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000463191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000463191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000463191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000463191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000465318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000465318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000465318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000465318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000465318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000466465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000467888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000467888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000467888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000467888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000467888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000468530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000468530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000468530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000468530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000468530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000468530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000475822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000475822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000475822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000475822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000483692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000483692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000483692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000483692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
ENST00000483692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007458.hg.1 |
NM_001430.4 |
Refseq |