Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023192.hg.1 |
TC07001540.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023192.hg.1 |
ENST00000307630.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023192.hg.1 |
ENST00000453207.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023192.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023187.hg.1 |
TC07001540.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023187.hg.1 |
ENST00000307630.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023187.hg.1 |
ENST00000536755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023187.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023194.hg.1 |
TC07001540.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023194.hg.1 |
ENST00000307630.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023194.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011570.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011570.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011570.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011570.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011570.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011570.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011570.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011570.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011570.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011570.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023188.hg.1 |
TC07001540.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023188.hg.1 |
ENST00000307630.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023188.hg.1 |
ENST00000453207.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023188.hg.1 |
ENST00000536755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023188.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023189.hg.1 |
TC07001540.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023189.hg.1 |
ENST00000307630.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023189.hg.1 |
ENST00000453207.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023189.hg.1 |
ENST00000536755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023189.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023186.hg.1 |
TC07001540.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023186.hg.1 |
ENST00000307630.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023186.hg.1 |
ENST00000453207.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023186.hg.1 |
ENST00000536755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023186.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023191.hg.1 |
TC07001540.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023191.hg.1 |
ENST00000307630.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023191.hg.1 |
ENST00000453207.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023191.hg.1 |
ENST00000536755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023191.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023190.hg.1 |
TC07001540.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023190.hg.1 |
ENST00000307630.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023190.hg.1 |
ENST00000453207.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023190.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182412.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182412.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182412.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182412.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182412.hg.1 |
TC0700011570.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700182411.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182411.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182411.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182411.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182411.hg.1 |
TC0700011570.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700182410.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182410.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182410.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182410.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182410.hg.1 |
TC0700011570.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700182413.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182413.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182413.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182413.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182413.hg.1 |
TC0700011570.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700011570.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011570.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011570.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011570.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011570.hg.1 |
ENST00000307630.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011570.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011570.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011570.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011570.hg.1 |
NM_012479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011570.hg.1 |
NM_012479.3 |
Refseq |