Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018203.hg.1 |
TC09001473.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018203.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018203.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374515.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374515.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374515.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374515.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374515.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374515.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000487892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000487892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000487892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
ENST00000487892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009438.hg.1 |
ENST00000454560.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009438.hg.1 |
ENST00000454560.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374515.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374515.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374515.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374515.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374515.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374515.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000487892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000487892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000487892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
ENST00000487892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_001244938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011177.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176020.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176020.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176020.hg.1 |
ENST00000374517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176020.hg.1 |
NM_003329.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176020.hg.1 |
TC0900011177.hg.1 |
Clariom D TC |