Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002806.3 | NP_002797.3 | 1 - 2 | 127 | 147 | |
XM_017021472.1 | XP_016876961.1 | 1 - 2 | 104 | 147 | ||
GenBank mRNA | AF006305.1 | - | 1 - 2 | 100 | 147 | |
AK307087.1 | - | 1 - 2 | 103 | 147 | ||
AK313670.1 | - | 1 - 2 | 95 | 147 | ||
BC005390.1 | - | 1 - 2 | 105 | 3996906 | 147 | |
BM788029.1 | - | 1 - 2 | 127 | 147 | ||
BT006843.1 | - | 1 - 2 | 85 | 147 | ||
CR456709.1 | - | 1 - 2 | 85 | 147 | ||
D78275.1 | - | 1 - 2 | 101 | 147 | ||
FJ224299.1 | - | 1 - 2 | 101 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons |