Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600842 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCKR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GCKR - glucokinase (hexokinase 4) regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001486.3 | 830 | Missense Mutation | CCG,CTG | P,L 446 | NP_001477.2 | |
XM_011532763.1 | 830 | Intron | XP_011531065.1 | |||
XM_017003796.1 | 830 | Missense Mutation | CCG,CTG | P,L 256 | XP_016859285.1 | |
XM_017003797.1 | 830 | Missense Mutation | CCG,CTG | P,L 256 | XP_016859286.1 |
Set Membership: |
HapMap Validated |