Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTGGGGAGATACTTTAGCAGGC[A/G]AGGGCTGTCAAGGAAAGTCTTATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608713 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP2R1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYP2R1 - cytochrome P450 family 2 subfamily R member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024514.4 | 5582 | Intron | NP_078790.2 | |||
XM_005252788.1 | 5582 | Intron | XP_005252845.1 | |||
XM_005252789.2 | 5582 | Intron | XP_005252846.1 | |||
XM_011519895.1 | 5582 | Intron | XP_011518197.1 | |||
XM_011519898.2 | 5582 | UTR 5 | XP_011518200.1 | |||
XM_017017190.1 | 5582 | Intron | XP_016872679.1 | |||
XM_017017191.1 | 5582 | Intron | XP_016872680.1 | |||
XM_017017192.1 | 5582 | Intron | XP_016872681.1 | |||
XM_017017193.1 | 5582 | Intron | XP_016872682.1 | |||
XM_017017194.1 | 5582 | Intron | XP_016872683.1 | |||
XM_017017195.1 | 5582 | Intron | XP_016872684.1 |
LOC107984314 - uncharacterized LOC107984314 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |