Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrtm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282102.1 | NP_001269031.1 | 2 | 1855 | ||
NM_178731.5 | NP_848846.3 | 2 | 1609 | |||
GenBank mRNA | AK038656.1 | - | 2 | 1686 |