Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slitrk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178740.4 | NP_848855.2 | 3 | 462 | ||
GenBank mRNA | AK032263.1 | - | 2 | 352 | ||
AK039763.1 | - | 3 | 403 | |||
AK046301.1 | - | 3 | 332 | |||
AK053065.1 | - | 3 | 463 | |||
BC117890.1 | - | 3 | 327 | 40096585 | ||
BC117891.1 | - | 2 | 260 | 40096591 |