Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178746.4 | NP_848861.1 | 2, 3 | 260 | ||
GenBank mRNA | AK043402.1 | - | 2, 3 | 248 | ||
AK044735.1 | - | 2, 3 | 250 | |||
BC052361.1 | - | 2, 3 | 260 | 6705789 |