Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lppr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178756.4 | NP_848871.1 | 3 | 335 | ||
GenBank mRNA | AK034251.1 | - | 3 | 520 | ||
AK043762.1 | - | 3 | 237 | |||
AK047614.1 | - | 3 | 265 | |||
AK053451.1 | - | 3 | 341 | |||
AK087439.1 | - | 3 | 486 | |||
AK087514.1 | - | 3 | 336 | |||
AK160897.1 | - | 3 | 462 | |||
AY345342.1 | - | 3 | 335 | |||
BC096762.1 | - | 3 | 311 | 30094664 |