Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6kc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178775.4 | NP_848890.3 | 9 | 1182 | ||
GenBank mRNA | AK044809.1 | - | 6 | 901 | ||
AK049003.1 | - | 2 | 130 | |||
AK149858.1 | - | 9 | 1147 |