Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178802.4 | NP_848917.2 | 3, 4 | 881 | ||
GenBank mRNA | AK028857.1 | - | 3, 4 | 770 | ||
AK047439.1 | - | 3, 4 | 881 | |||
AK050080.1 | - | 3, 4 | 793 | |||
AK079982.1 | - | 3 | 963 | |||
BC057094.1 | - | 770 | 6418691 | |||
BC118954.1 | - | 3, 4 | 636 | |||
BC118955.1 | - | 3, 4 | 636 |