Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178873.3 | NP_849204.1 | 1 | 502 | ||
GenBank mRNA | AK028568.1 | - | 1 | 503 | ||
AK161225.1 | - | 1 | 505 | |||
BC069944.1 | - | 1 | 250 | 30446117 |