Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007245.2 | NP_001007246.1 | 4 | 697 | ||
NM_001197079.1 | NP_001184008.1 | 3 | 587 | |||
NM_001197080.1 | NP_001184009.1 | 3 | 422 | |||
NM_001550.3 | NP_001541.2 | 3 | 488 | |||
NR_120333.1 | 3 | 408 | ||||
XM_011516142.2 | XP_011514444.1 | 3 | 483 | |||
XM_011516143.2 | XP_011514445.1 | 3 | 668 | |||
XM_017012151.1 | XP_016867640.1 | 3 | 735 | |||
GenBank mRNA | AK225222.1 | - | 3 | 461 | ||
AK297509.1 | - | 3 | 233 | |||
AK298894.1 | - | 3 | 228 | |||
AK308066.1 | - | 4 | 672 | |||
AK313211.1 | - | 4 | 697 | |||
AK315967.1 | - | 3 | 420 | |||
AU280361.1 | - | 3 | 485 | |||
BC001272.1 | - | 3 | 441 | 3458309 | ||
BT019889.1 | - | 3 | 227 | |||
BT019890.1 | - | 3 | 227 | |||
DC352698.1 | - | 3 | 488 |