Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178726.3 | NP_848841.2 | 4 | 1712 | ||
GenBank mRNA | AK028275.1 | - | 4 | 754 | ||
AK032529.1 | - | 4 | 1770 | |||
AK035912.1 | - | 4 | 1715 | |||
AK045724.1 | - | 4 | 1753 | |||
AK131646.1 | - | 4 | 1699 | |||
AK142789.1 | - | 4 | 1731 | |||
AK147389.1 | - | 4 | 1713 | |||
AK147876.1 | - | 4 | 1749 | |||
AK165160.1 | - | 4 | 1747 | |||
AK220523.1 | - | 4 | 1717 | |||
AY184801.1 | - | 4 | 1300 | |||
BC096031.1 | - | 4 | 1712 | 6837066 |