Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AW551984 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199556.1 | NP_001186485.1 | 5 | 503 | ||
NM_178737.5 | NP_848852.1 | 5 | 493 | |||
GenBank mRNA | AK028282.1 | - | 5 | 467 | ||
AK049098.1 | - | 5 | 494 | |||
AK082321.1 | - | 5 | 476 | |||
BC061211.1 | - | 5 | 405 | 30250574 |